(R)-(+)-3-Phenylcyclohexanone ≥97.0%

Supplier: TCI America

P1499-1G P1499-100MG
TCP1499-1GEA 764.72 CAD
TCP1499-1G TCP1499-100MG
(R)-(+)-3-Phenylcyclohexanone ≥97.0%
(R)-(+)-3-Phenylcyclohexanone

CAS Number: 34993-51-6
Molecular Formula: C12H14O
Molecular Weight: 174.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.05
Specific rotation [a]20/D: 11 deg (neat)
Storage Temperature: 0-10°C

Formula: C₁₂H₁₄O
Density: 1.05 (20 °C)
CAS Number: 34993-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=823bc4d764e607f49ede3287be526775186ae29c&vid=823bc4d764e607f49ede3287be526775186ae29c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR