N-Phenyl-9-anthramine ≥98.0%

Supplier: TCI America

P1496-1G P1496-5G
TCP1496-1GEA 180.12 CAD
TCP1496-1G TCP1496-5G
N-Phenyl-9-anthramine ≥98.0%
N-Phenyl-9-anthramine

CAS Number: 15424-38-1
Molecular Formula: C20H15N
Molecular Weight: 269.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 207


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 9-Anilinoanthracene

Formula: C₂₀H₁₅N
Melting Pt: 207 °C
MDL Number: MFCD03093237
CAS Number: 15424-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f92bc1de1738f41095ad0cf7461debb39a1ef05&vid=5f92bc1de1738f41095ad0cf7461debb39a1ef05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR