9-Phenylcarbazole ≥98.0%
Supplier: TCI America
P1492-5G
P1492-1G
P1492-25G
TCP1492-5GEA
75.84
CAD
TCP1492-5G
TCP1492-1G
TCP1492-25G
9-Phenylcarbazole ≥98.0%
9-Phenylcarbazole
CAS Number: 1150-62-5
Molecular Formula: C18H13N
Molecular Weight: 243.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 88
Formula:
C₁₈H₁₃N Melting Pt: 88 °C |
MDL Number:
MFCD00004965 CAS Number: 1150-62-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...