(R)-Propylene carbonate ≥98.0%
Supplier: TCI America
P1485-5G
P1485-25G
TCP1485-5GEA
79
CAD
TCP1485-5G
TCP1485-25G
(R)-Propylene carbonate ≥98.0%
(R)-Propylene carbonate
CAS Number: 16606-55-6
MDL Number: MFCD00798265
Molecular Formula: C4H6O3
Molecular Weight: 102.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 132
Specific Gravity (20/20): 1.21
Specific rotation [a]20/D: 2 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-4-Methyl-1,3-dioxolan-2-one
Formula:
C₄H₆O₃ Density: 1.21 (20 °C) Flash Pt: 132 °C |
MDL Number:
MFCD00798265 CAS Number: 16606-55-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...