Potassium allyltrifluoroborate ≥95.0% (by gravimetric analysis)
Supplier: TCI America
P1480-1G
P1480-5G
TCP1480-1GEA
102.7
CAD
TCP1480-1G
TCP1480-5G
Potassium allyltrifluoroborate ≥95.0% (by gravimetric analysis)
Potassium allyltrifluoroborate
CAS Number: 233664-53-4
Molecular Formula: C3H6BF3
Molecular Weight: 147.98
Purity/Analysis Method: >95.0% (W)
Form: Crystal
Color: White
Formula:
C₃H₅BF₃K |
MDL Number:
MFCD07368222 CAS Number: 233664-53-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...