Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥98.0% (by HPLC)

Supplier: TCI America

P1476-5G P1476-25G
TCP1476-5GEA 243 CAD
TCP1476-5G TCP1476-25G
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥98.0% (by HPLC)
Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-beta-D-glucopyranoside

CAS Number: 23661-28-1
MDL Number: MFCD00135159
Molecular Formula: C20H24O9S
Molecular Weight: 440.46
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 118
Storage Temperature: <0°C

Formula: C₂₀H₂₄O₉S
Melting Pt: 118 °C
MDL Number: MFCD00135159
CAS Number: 23661-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22afc64a3eadfe93e2c7b528109f759e4add53ab&vid=22afc64a3eadfe93e2c7b528109f759e4add53ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR