2-(4-Pyridyl)-2-propanol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(4-Pyridyl)isopropyl Alcohol

P1471-5G P1471-25G
TCP1471-5GEA 88.48 CAD
TCP1471-5G TCP1471-25G
2-(4-Pyridyl)-2-propanol ≥97.0% (by GC, titration analysis)
2-(4-Pyridyl)-2-propanol

CAS Number: 15031-78-4
MDL Number: MFCD00071746
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 140
Melting point (°C): 136


Other Product Names / Synonyms: 2-(4-Pyridyl)isopropyl Alcohol

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 140 °C
Melting Pt: 136 °C
MDL Number: MFCD00071746
CAS Number: 15031-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e1c192a82dcfa8c894df7a1b78be91668ed9ddf&vid=7e1c192a82dcfa8c894df7a1b78be91668ed9ddf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR