2-(4-Piperidyl)-2-propanol ≥98.0%

Supplier: TCI America

Synonyms: 2-(4-Piperidyl)isopropyl Alcohol

P1470-1G P1470-5G P1470-25G
TCP1470-1GEA 64.78 CAD
TCP1470-1G TCP1470-5G TCP1470-25G
2-(4-Piperidyl)-2-propanol ≥98.0%
2-(4-Piperidyl)-2-propanol

CAS Number: 22990-34-7
MDL Number: MFCD06797159
Molecular Formula: C8H17NO
Molecular Weight: 143.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 136


Other Product Names / Synonyms: 2-(4-Piperidyl)isopropyl Alcohol

Formula: C₈H₁₇NO
Melting Pt: 136 °C
MDL Number: MFCD06797159
CAS Number: 22990-34-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75888c1605206cf4bc5bb47ef6b614e8641345b7&vid=75888c1605206cf4bc5bb47ef6b614e8641345b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR