4-Propargylthiomorpholine-1,1-dioxide ≥98.0%

Supplier: TCI America

P1469-25G P1469-5G
TCP1469-25GEA 773.57 CAD
TCP1469-25G TCP1469-5G
4-Propargylthiomorpholine-1,1-dioxide ≥98.0%
4-Propargylthiomorpholine-1,1-dioxide

CAS Number: 10442-03-2
MDL Number: MFCD00085157
Molecular Formula: C7H11NO2S
Molecular Weight: 173.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 116


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-(2-Propynyl)thiomorpholine 1,1-Dioxide

Formula: C₇H₁₁NO₂S
Melting Pt: 116 °C
MDL Number: MFCD00085157
CAS Number: 10442-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55cba67b05b36f847d921bb68ed4bf7d4c88fef4&vid=55cba67b05b36f847d921bb68ed4bf7d4c88fef4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR