2,3,4,5,6-Pentabromobenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (2,3,4,5,6-Pentabromophenyl)methanol, Pentabromobenzyl alcohol, 2,3,4,5,6-Pentabromobenzylalcohol

P1457-1G P1457-5G
TCP1457-1GEA 56.88 CAD
TCP1457-1G TCP1457-5G
2,3,4,5,6-Pentabromobenzyl alcohol ≥98.0%
2,3,4,5,6-Pentabromobenzyl alcohol

CAS Number: 79415-41-1
MDL Number: MFCD00192377
Molecular Formula: C7H3Br5O
Molecular Weight: 502.62
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow

Formula: C₇H₃Br₅O
MW: 502.62 g/mol
MDL Number: MFCD00192377
CAS Number: 79415-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81b16b0445733c6effd204ec769f77ed6b058b7d&vid=81b16b0445733c6effd204ec769f77ed6b058b7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR