4H-Pyran-4-one ≥98.0%
Supplier: TCI America
P1450-1G
P1450-5G
TCP1450-1GEA
304.15
CAD
TCP1450-1G
TCP1450-5G
4H-Pyran-4-one ≥98.0%
4H-Pyran-4-one
CAS Number: 108-97-4
MDL Number: MFCD00006576
Molecular Formula: C5H4O2
Molecular Weight: 96.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 215
Melting point (°C): 32
Flash Point (°C): 101
Other Product Names / Synonyms: gamma-Pyrone
Formula:
C₅H₄O₂ Boiling Pt: 215 °C Melting Pt: 32 °C Flash Pt: 101 °C |
MDL Number:
MFCD00006576 CAS Number: 108-97-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...