4-Phenyl-1-cyclohexene ≥96.0%

Supplier: TCI America

Synonyms: 3-Cyclohexen-1-ylbenzene

P1445-1G P1445-5G
TCP1445-1GEA 383.94 CAD
TCP1445-1G TCP1445-5G
4-Phenyl-1-cyclohexene ≥96.0%
4-Phenyl-1-cyclohexene

CAS Number: 4994-16-5
MDL Number: MFCD00049188
Molecular Formula: C12H14
Molecular Weight: 158.24
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 235
Freezing point (°C): -14
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 3-Cyclohexen-1-ylbenzene

Formula: C₁₂H₁₄
Boiling Pt: 235 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00049188
CAS Number: 4994-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d16450906a9684fe14268dbeedff1fd1f16b0ff&vid=9d16450906a9684fe14268dbeedff1fd1f16b0ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR