Dimethyl-2,6-pyridinedicarboxylate ≥98.0%

Supplier: TCI America

Synonyms: Dimethyl pyridine-2,6-dicarboxylate

P1444-25G P1444-5G
TCP1444-25GEA 134.3 CAD
TCP1444-25G TCP1444-5G
Dimethyl-2,6-pyridinedicarboxylate ≥98.0%
Dimethyl 2,6-Pyridinedicarboxylate

CAS Number: 5453-67-8
MDL Number: MFCD00134493
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 124


Other Product Names / Synonyms: Dimethyl Dipicolinate, Dipicolinic Acid Dimethyl Ester, 2,6-Pyridinedicarboxylic Acid Dimethyl Ester

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 124 °C
MDL Number: MFCD00134493
CAS Number: 5453-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bc5597b35c10c44026fd47c227688af537da568&vid=8bc5597b35c10c44026fd47c227688af537da568&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR