4-Phenylethynylphthalic anhydride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-(Phenylethynyl)isobenzofuran-1,3-dione

P1437-25G P1437-5G
TCP1437-25GEA 578.28 CAD
TCP1437-25G TCP1437-5G
4-Phenylethynylphthalic anhydride ≥98.0% (by GC, titration analysis)
4-Phenylethynylphthalic anhydride

CAS Number: 119389-05-8
MDL Number: MFCD01862632
Molecular Formula: C16H8O3
Molecular Weight: 248.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 152

Formula: C₁₆H₈O₃
MW: 248.24 g/mol
Melting Pt: 152 °C
MDL Number: MFCD01862632
CAS Number: 119389-05-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b73b9e426f90bfef2097656a45a51af031b39cd0&vid=b73b9e426f90bfef2097656a45a51af031b39cd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR