6-[2-Propyl-4-(4-pyridylazo)phenoxy]hexanoic acid ≥98.0%

Supplier: TCI America

P1436-1G P1436-5G
TCP1436-1GEA 181.86 CAD
TCP1436-1G TCP1436-5G
6-[2-Propyl-4-(4-pyridylazo)phenoxy]hexanoic acid ≥98.0%
6-[2-Propyl-4-(4-pyridylazo)phenoxy]hexanoic acid

CAS Number: 312694-02-3
Molecular Formula: C20H25N3O3
Molecular Weight: 355.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 173

Formula: C₂₀H₂₅N₃O₃
MW: 355.44 g/mol
Melting Pt: 173 °C
CAS Number: 312694-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0a57660682307ef8d6c2533750ce213169e7d40&vid=c0a57660682307ef8d6c2533750ce213169e7d40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR