Diethyl-1,4-phenylenediacetate ≥98.0%

Supplier: TCI America

Synonyms: Diethyl 2,2-(1,4-phenylene)diacetate, 1,4-Phenylenediacetic Acid Diethyl Ester

P1434-25G P1434-5G
TCP1434-25GEA 600.08 CAD
TCP1434-25G TCP1434-5G
Diethyl-1,4-phenylenediacetate ≥98.0%
Diethyl-1,4-phenylenediacetate

CAS Number: 36076-26-3
MDL Number: MFCD00032366
Molecular Formula: C14H18O4
Molecular Weight: 250.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 140
Melting point (°C): 59


Other Product Names / Synonyms: 1,4-Phenylenediacetic Acid Diethyl Ester

Formula: C₁₄H₁₈O₄
MW: 250.29 g/mol
Boiling Pt: 140 °C
Melting Pt: 59 °C
MDL Number: MFCD00032366
CAS Number: 36076-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f356d3a342570e8d900fbf37de5def4f2aae3fbe&vid=f356d3a342570e8d900fbf37de5def4f2aae3fbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR