2-Phenyl-1,3-dioxan-5-ol ≥90.0% (by NMR)

Supplier: TCI America

P1422-1G P1422-5G P1422-25G
TCP1422-1GEA 274.92 CAD
TCP1422-1G TCP1422-5G TCP1422-25G
2-Phenyl-1,3-dioxan-5-ol ≥90.0% (by NMR)
2-Phenyl-1,3-dioxan-5-ol

CAS Number: 1708-40-3
MDL Number: MFCD00047504
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >90.0% (NMR)
Form: Crystal
Color: White
Melting point (°C): 84
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,3-O-Benzylideneglycerol, 5-Hydroxy-2-phenyl-1,3-dioxane

Formula: C₁₀H₁₂O₃
Melting Pt: 84 °C
MDL Number: MFCD00047504
CAS Number: 1708-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364205b7e5c33ebe11d36eacf876077669c32ea1&vid=364205b7e5c33ebe11d36eacf876077669c32ea1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR