Tris(2,4-di-tert-butylphenyl)phosphite ≥98.0%
Supplier: TCI America
P1421-25G
P1421-500G
P1421-100G
TCP1421-25GEA
55.96
CAD
TCP1421-25G
TCP1421-500G
TCP1421-100G
Tris(2,4-di-tert-butylphenyl)phosphite ≥98.0%
Tris(2,4-di-tert-butylphenyl)phosphite
CAS Number: 31570-04-4
MDL Number: MFCD00072706
Molecular Formula: C42H63O3P
Molecular Weight: 646.94
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 187
Flash Point (°C): 150
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Phosphorous Acid Tris(2,4-di-tert-butylphenyl) Ester
Formula:
C₄₂H₆₃O₃P Melting Pt: 187 °C Flash Pt: 150 °C |
MDL Number:
MFCD00072706 CAS Number: 31570-04-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...