1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate)

Supplier: TCI America

Synonyms: 1,1'-(p-Xylylene)bis(4,4'-bipyridinium) Bis(hexafluorophosphate)

P1407-1G P1407-100MG
TCP1407-1GEA 1360.38 CAD
TCP1407-1G TCP1407-100MG
1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate)
1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)bis(hexafluorophosphate)

CAS Number: 108861-20-7
MDL Number: MFCD02063876
Molecular Formula: C28H24F12N4P2
Molecular Weight: 706.46
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: 1,1'-(p-Xylylene)bis(4,4'-bipyridinium) Bis(hexafluorophosphate)

Formula: C₂₈H₂₄F₁₂N₄P₂
MW: 706.46 g/mol
MDL Number: MFCD02063876
CAS Number: 108861-20-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7818b1e65b924103dbbf951becae440588dc0c8f&vid=7818b1e65b924103dbbf951becae440588dc0c8f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR