4-Phenylpyridine-N-oxide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
P1406-5G
P1406-25G
TCP1406-5GEA
398.16
CAD
TCP1406-5G
TCP1406-25G
4-Phenylpyridine-N-oxide ≥98.0% (by HPLC, titration analysis)
4-Phenylpyridine N-oxide
CAS Number: 1131-61-9
MDL Number: MFCD00006208
Molecular Formula: C11H9NO
Molecular Weight: 171.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 155
Formula:
C₁₁H₉NO Melting Pt: 155 °C |
MDL Number:
MFCD00006208 CAS Number: 1131-61-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...