1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)dibromide ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
1,1'-(p-Xylylene)bis(4,4'-bipyridinium) Dibromide
P1405-100MG
P1405-1G
TCP1405-100MGEA
102.7
CAD
TCP1405-100MG
TCP1405-1G
1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)dibromide ≥98.0% (by HPLC)
1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium)dibromide
CAS Number: 106867-97-4
MDL Number: MFCD03844804
Molecular Formula: C28H24Br2N4
Molecular Weight: 576.34
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Other Product Names / Synonyms: 1,1'-(p-Xylylene)bis(4,4'-bipyridinium) Dibromide
Formula:
C₂₈H₂₄Br₂N₄ MW: 576.34 g/mol |
MDL Number:
MFCD03844804 CAS Number: 106867-97-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...