2-Pyridyl tribromomethyl sulfone ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

P1377-5G P1377-25G
TCP1377-005GEA 178.54 CAD
TCP1377-005G TCP1377-025G
2-Pyridyl tribromomethyl sulfone ≥98.0% (by HPLC, total nitrogen)
2-Pyridyl tribromomethyl sulfone

CAS Number: 59626-33-4
MDL Number: MFCD02093493
Molecular Formula: C6H4Br3NO2S
Molecular Weight: 393.88
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Flash Point (°C): 176


Other Product Names / Synonyms: 2-(Tribromomethanesulfonyl)pyridine

Formula: C₆H₄Br₃NO₂S
Flash Pt: 176 °C
MDL Number: MFCD02093493
CAS Number: 59626-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8544226d8b1e8bcad123b7973bdee23258ca0adf&vid=8544226d8b1e8bcad123b7973bdee23258ca0adf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR