4-Pyridineethanethiol hydrochloride ≥97.0%
Supplier: TCI America
Synonyms:
4-(2-Mercaptoethyl)pyridine Hydrochloride, 2-(Pyridin-4-yl)ethanethiol hydrochloride, 2-(4-Pyridyl)ethanethiol Hydrochloride
P1373-25G
P1373-5G
TCP1373-025GEA
818.12
CAD
TCP1373-025G
TCP1373-5G
4-Pyridineethanethiol hydrochloride ≥97.0%
4-Pyridineethanethiol hydrochloride
CAS Number: 6298-11-9
MDL Number: MFCD00956047
Molecular Formula: C7H9NS
Molecular Weight: 175.67
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Other Product Names / Synonyms: 4-(2-Mercaptoethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanethiol Hydrochloride
Formula:
C₇H₉NS MW: 175.67 g/mol |
MDL Number:
MFCD00956047 CAS Number: 6298-11-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...