Sodium-4-aminobenzoate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Aminobenzoic Acid Sodium Salt, Sodium 4-aminobenzoate

A0276-500G A0276-25G
TCA0276-500GEA 567.22 CAD
TCA0276-500G TCA0276-025G
Sodium-4-aminobenzoate ≥98.0% (by HPLC, titration analysis)
Sodium-4-aminobenzoate

CAS Number: 555-06-6
MDL Number: MFCD00064395
Molecular Formula: C7H7NO2
Molecular Weight: 159.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Aminobenzoic Acid Sodium Salt

Formula: C₇H₇NO₂
MW: 159.12 g/mol
MDL Number: MFCD00064395
CAS Number: 555-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08080e322890d37cc68fe2608a020efc88e7cef1&vid=08080e322890d37cc68fe2608a020efc88e7cef1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR