Pentamethylenebis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) ≥97.0% (by HPLC)

Supplier: TCI America

P1364-100MG P1364-10MG
TCP1364-100MGEA 1393.4 CAD
TCP1364-100MG TCP1364-010MG
Pentamethylenebis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) ≥97.0% (by HPLC)
Pentamethylenebis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II)

[Reagent for application of the exciton chirality method]
CAS Number: 210769-64-5
MDL Number: MFCD02093489
Molecular Formula: C95H64N8O4Zn2
Molecular Weight: 1512.37
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: Deep Purple
Storage Temperature: 0-10°C


Product Categories: Bioscience, Porphyrins, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 1,5-Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoyloxy]pentane Dizinc(II), Pentamethylene Bis[4-(10,15,20-triphenylporphin-5-yl)benzoate]dizinc(II)

Formula: C₉₅H₆₄N₈O₄Zn₂
MDL Number: MFCD02093489
CAS Number: 210769-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f3f992caafd315b943d3790b452e1b26a37f717&vid=5f3f992caafd315b943d3790b452e1b26a37f717&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR