2-Phenylthiomethylbenzoic acid
Supplier: TCI America
P1360-25G
P1360-5G
TCP1360-025GEA
658.86
CAD
TCP1360-025G
TCP1360-005G
2-Phenylthiomethylbenzoic acid
2-Phenylthiomethylbenzoic acid
CAS Number: 1699-03-2
MDL Number: MFCD00089444
Molecular Formula: C14H12O2S
Molecular Weight: 244.31
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 111
Other Product Names / Synonyms: 2-Phenylmercaptomethylbenzoic Acid
Formula:
C₁₄H₁₂O₂S Melting Pt: 111 °C |
MDL Number:
MFCD00089444 CAS Number: 1699-03-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...