Di-tert-amyldicarbonate ≥95.0%

Supplier: TCI America

P1356-5G P1356-25G
TCP1356-005GEA 274.02 CAD
TCP1356-005G TCP1356-025G
Di-tert-amyldicarbonate ≥95.0%
Di-tert-amyldicarbonate

CAS Number: 68835-89-2
MDL Number: MFCD00038320
Molecular Formula: C12H22O5
Molecular Weight: 246.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 71
Flash Point (°C): 104
Specific Gravity (20/20): 1.01
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Di-tert-amyl Pyrocarbonate, Pyrocarbonic Acid Di-tert-amyl Ester

Formula: C₁₂H₂₂O₅
Boiling Pt: 71 °C
Density: 1.01 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00038320
CAS Number: 68835-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42285838adcc27a658ec60f6bb4ab2e977ce9a95&vid=42285838adcc27a658ec60f6bb4ab2e977ce9a95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR