Chromium(III) picolinate ≥96.0% (by titrimetric analysis)

Supplier: TCI America

P1347-25G P1347-5G
TCP1347-025GEA 330.54 CAD
TCP1347-025G TCP1347-5G
Chromium(III) picolinate ≥96.0% (by titrimetric analysis)
Chromium(III) picolinate

CAS Number: 14639-25-9
MDL Number: MFCD00068715
Molecular Formula: C6H5NO2
Molecular Weight: 418.31
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: Slightly Pale Purple


Product Categories: Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Chromium(III) Picolinate, Picolinic Acid Chromium(III) Salt, Pyridine-2-carboxylic Acid Chromium(III) Salt

Formula: C₁₈H₁₂CrN₃O₆
MDL Number: MFCD00068715
CAS Number: 14639-25-9

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR