Methyl-5-phenylvalerate ≥98.0%

Supplier: TCI America

P1324-5G P1324-25G
TCP1324-005GEA 213.3 CAD
TCP1324-005G TCP1324-25G
Methyl-5-phenylvalerate ≥98.0%
Methyl-5-phenylvalerate

CAS Number: 20620-59-1
MDL Number: MFCD00025896
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 173
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: 5-Phenylvaleric Acid Methyl Ester

Formula: C₁₂H₁₆O₂
Boiling Pt: 173 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00025896
CAS Number: 20620-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e296af347d86aeb85d1b404114b122aaf70834a&vid=8e296af347d86aeb85d1b404114b122aaf70834a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR