Ethyl phenylsulfonylacetate ≥98.0%

Supplier: TCI America

P1306-25G P1306-5G
TCP1306-025GEA 257.54 CAD
TCP1306-025G TCP1306-005G
Ethyl phenylsulfonylacetate ≥98.0%
Ethyl phenylsulfonylacetate

CAS Number: 7605-30-3
MDL Number: MFCD00025041
Molecular Formula: C10H12O4S
Molecular Weight: 228.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 44


Other Product Names / Synonyms: Phenylsulfonylacetic Acid Ethyl Ester

Formula: C₁₀H₁₂O₄S
Melting Pt: 44 °C
MDL Number: MFCD00025041
CAS Number: 7605-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f20a8540df91e6865a827da18eb6fc633cca5d1&vid=6f20a8540df91e6865a827da18eb6fc633cca5d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR