Dimethyl-2,3-pyridinedicarboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1303-25G P1303-5G
TCP1303-25GEA 216.46 CAD
TCP1303-25G TCP1303-005G
Dimethyl-2,3-pyridinedicarboxylate ≥98.0% (by GC, titration analysis)
Dimethyl 2,3-Pyridinedicarboxylate

CAS Number: 605-38-9
MDL Number: MFCD00186549
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 56


Other Product Names / Synonyms: Dimethyl Quinolinate, 2,3-Pyridinedicarboxylic Acid Dimethyl Ester, Quinolinic Acid Dimethyl Ester

Formula: C₉H₉NO₄
Melting Pt: 56 °C
MDL Number: MFCD00186549
CAS Number: 605-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e551dc20e255bd0716237fcf18eb1634a3e30e0a&vid=e551dc20e255bd0716237fcf18eb1634a3e30e0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR