[Hydroxy(methanesulfonyloxy)iodo]benzene ≥98.0%

Supplier: TCI America

Synonyms: Phenyliodosohydroxy Methanesulphonate, [Hydroxy(mesyloxy)iodo]benzene, Phenyliodosohydroxy Methanesulfonate

P1298-5G P1298-25G
TCP1298-005GEA 125.61 CAD
TCP1298-005G TCP1298-25G
[Hydroxy(methanesulfonyloxy)iodo]benzene ≥98.0%
[Hydroxy(methanesulfonyloxy)iodo]benzene

CAS Number: 105551-42-6
MDL Number: MFCD01321184
Molecular Formula: C7H9IO4S
Molecular Weight: 316.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 122


Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds
Other Product Names / Synonyms: [Hydroxy(mesyloxy)iodo]benzene, Phenyliodosohydroxy Methanesulfonate

Formula: C₇H₉IO₄S
Melting Pt: 122 °C
MDL Number: MFCD01321184
CAS Number: 105551-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e75e61b9e4e931346166eaabad9a7e1007b5f01&vid=9e75e61b9e4e931346166eaabad9a7e1007b5f01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR