3-Phenyl-2,4-pentanedione ≥98.0%

Supplier: TCI America

P1297-5G P1297-1G
TCP1297-005GEA 551.42 CAD
TCP1297-005G TCP1297-1G
3-Phenyl-2,4-pentanedione ≥98.0%
3-Phenyl-2,4-pentanedione

CAS Number: 5910-25-8
MDL Number: MFCD00048223
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 134
Melting point (°C): 56


Product Categories: Synthetic Organic Chemistry, Oxidation, Ligands
Other Product Names / Synonyms: 3-Phenylacetylacetone

Formula: C₁₁H₁₂O₂
Boiling Pt: 134 °C
Melting Pt: 56 °C
MDL Number: MFCD00048223
CAS Number: 5910-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=128fbfdac742a697aa02242aa6fe1c41a73bce24&vid=128fbfdac742a697aa02242aa6fe1c41a73bce24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR