4-Pyridineethanol hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 4-(2-Hydroxyethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanol Hydrochloride

P1293-25G P1293-5G
TCP1293-25GEA 1576.84 CAD
TCP1293-25G TCP1293-005G
4-Pyridineethanol hydrochloride ≥98.0%
4-Pyridineethanol hydrochloride

CAS Number: 383177-54-6
MDL Number: MFCD01321183
Molecular Formula: C7H9NO
Molecular Weight: 159.61
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 151


Other Product Names / Synonyms: 4-(2-Hydroxyethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanol Hydrochloride

Formula: C₇H₉NO
MW: 159.61 g/mol
Melting Pt: 151 °C
MDL Number: MFCD01321183
CAS Number: 383177-54-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5d22080b45151741fb716c0ff94ab1338b1ae38&vid=d5d22080b45151741fb716c0ff94ab1338b1ae38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR