Dibenzyl dicarbonate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dibenzyl pyrocarbonate

P1281-5G P1281-25G
TCP1281-005GEA 196.3 CAD
TCP1281-005G TCP1281-25G
Dibenzyl dicarbonate ≥97.0% (by titrimetric analysis)
Dibenzyl dicarbonate

CAS Number: 31139-36-3
MDL Number: MFCD00043124
Molecular Formula: C16H14O5
Molecular Weight: 286.28
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Flash Point (°C): 110
Freezing point (°C): 23


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Peptide Synthesis, Bioscience, Polyamine Synthesis Reagents
Other Product Names / Synonyms: Dibenzyl Pyrocarbonate, Pyrocarbonic Acid Dibenzyl Ester

Formula: C₁₆H₁₄O₅
MW: 286.28 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00043124
CAS Number: 31139-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=044c13831c21cfddbdbccf1e01a9ee643ad4a2f9&vid=044c13831c21cfddbdbccf1e01a9ee643ad4a2f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR