2-Adamantanone oxime ≥98.0%

Supplier: TCI America

A2110-25G A2110-5G
TCA2110-25GEA 472.74 CAD
TCA2110-25G TCA2110-5G
2-Adamantanone oxime ≥98.0%
2-Adamantanone oxime

CAS Number: 4500-12-3
MDL Number: MFCD00078273
Molecular Formula: C10H15NO
Molecular Weight: 165.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 168

Formula: C₁₀H₁₅NO
Melting Pt: 168 °C
MDL Number: MFCD00078273
CAS Number: 4500-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3187b9f9dcfc2a99aa3cadf692474a864182867&vid=b3187b9f9dcfc2a99aa3cadf692474a864182867&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR