9-Phenylxanthen-9-ol
Supplier: TCI America
P1269-5G
P1269-25G
TCP1269-005GEA
455.04
CAD
TCP1269-005G
TCP1269-025G
9-Phenylxanthen-9-ol
9-Phenylxanthen-9-ol
CAS Number: 596-38-3
MDL Number: MFCD00005058
Molecular Formula: C19H14O2
Molecular Weight: 274.32
Form: Crystal
Color: White
Melting point (°C): 162
Formula:
C₁₉H₁₄O₂ Melting Pt: 162 °C |
MDL Number:
MFCD00005058 CAS Number: 596-38-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...