Methyl-2-pyridylacetate ≥97.0%

Supplier: TCI America

Synonyms: 2-Pyridineacetic Acid Methyl Ester, Methyl 2-Pyridineacetate, Methyl 2-(pyridin-2-yl)acetate, 2-Pyridylacetic Acid Methyl Ester

P1268-25G P1268-5G
TCP1268-025GEA 327.52 CAD
TCP1268-025G TCP1268-5G
Methyl-2-pyridylacetate ≥97.0%
Methyl-2-pyridylacetate

CAS Number: 1658-42-0
MDL Number: MFCD00006358
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 137
Flash Point (°C): 113
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: Methyl 2-Pyridineacetate, 2-Pyridineacetic Acid Methyl Ester, 2-Pyridylacetic Acid Methyl Ester

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Boiling Pt: 137 °C
Density: 1.13 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00006358
CAS Number: 1658-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbff785b19a78db88ce0b3faa0d457213a4916a9&vid=bbff785b19a78db88ce0b3faa0d457213a4916a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR