(2-Methoxyethyl)benzene ≥98.0%

Supplier: TCI America

P1263-25ML P1263-500ML
TCP1263-025MLEA 61.62 CAD
TCP1263-025ML TCP1263-500ML
(2-Methoxyethyl)benzene ≥98.0%
(2-Methoxyethyl)benzene

CAS Number: 3558-60-9
MDL Number: MFCD00025912
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 190
Flash Point (°C): 80
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Methyl Phenethyl Ether

Formula: (C₆H₅)CH₂CH₂OCH₃
Boiling Pt: 190 °C
Density: 0.95 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00025912
CAS Number: 3558-60-9
EINECS: 222-619-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b656e066bcae6e71ffdb80ddab7ea618a27b56b6&vid=b656e066bcae6e71ffdb80ddab7ea618a27b56b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR