Ethyl phenylcyanoacetate ≥95.0%
Supplier: TCI America
P1255-5G
P1255-25G
TCP1255-005GEA
74.26
CAD
TCP1255-005G
TCP1255-025G
Ethyl phenylcyanoacetate ≥95.0%
Ethyl phenylcyanoacetate
CAS Number: 4553-07-5
MDL Number: MFCD00001866
Molecular Formula: C11H11NO2
Molecular Weight: 189.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 275
Flash Point (°C): 110
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: Phenylcyanoacetic Acid Ethyl Ester
Formula:
C₁₁H₁₁NO₂ Boiling Pt: 275 °C Density: 1.09 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00001866 CAS Number: 4553-07-5 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...