N-(Phenylthio)phthalimide ≥98.0%

Supplier: TCI America

Synonyms: 2-(Phenylthio)isoindoline-1,3-dione

P1254-5G P1254-25G
TCP1254-005GEA 175.22 CAD
TCP1254-005G TCP1254-025G
N-(Phenylthio)phthalimide ≥98.0%
N-(Phenylthio)phthalimide

CAS Number: 14204-27-4
MDL Number: MFCD00192396
Molecular Formula: C14H9NO2S
Molecular Weight: 255.29
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 162

Formula: C₁₄H₉NO₂S
Melting Pt: 162 °C
MDL Number: MFCD00192396
CAS Number: 14204-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed7d902d44f35408b41856714cfea25fdda9de1e&vid=ed7d902d44f35408b41856714cfea25fdda9de1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR