5-Allyl-3-methoxysalicylaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 5-Allyl-2-hydroxy-3-methoxybenzaldehyde

A2104-1G A2104-5G
TCA2104-1GEA 72.68 CAD
TCA2104-1G TCA2104-5G
5-Allyl-3-methoxysalicylaldehyde ≥98.0%
5-Allyl-3-methoxysalicylaldehyde

CAS Number: 22934-51-6
MDL Number: MFCD06408022
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 167
Melting point (°C): 48


Other Product Names / Synonyms: 5-Allyl-2-hydroxy-3-methoxybenzaldehyde

Formula: C₁₁H₁₂O₃
Boiling Pt: 167 °C
Melting Pt: 48 °C
MDL Number: MFCD06408022
CAS Number: 22934-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b1f0fb8c7cdd47e77fbd22db58f22d2770010dd&vid=0b1f0fb8c7cdd47e77fbd22db58f22d2770010dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR