Phenyl trimethylsilylmethyl sulfone ≥95.0%

Supplier: TCI America

Synonyms: (Phenylsulphonylmethyl)trimethylsilane, (Phenylsulfonylmethyl)trimethylsilane

P1250-1G P1250-5G
TCP1250-1GEA 386.78 CAD
TCP1250-1G TCP1250-005G
Phenyl trimethylsilylmethyl sulfone ≥95.0%
Phenyl trimethylsilylmethyl sulfone

CAS Number: 17872-92-3
MDL Number: MFCD00012364
Molecular Formula: C10H16O2SSi
Molecular Weight: 228.38
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 110
Freezing point (°C): 25


Product Categories: Synthetic Organic Chemistry, Sulfones, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (Phenylsulfonylmethyl)trimethylsilane

Formula: C₁₀H₁₆O₂SSi
MW: 228.38 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00012364
CAS Number: 17872-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05a6dd2d7ca5ffe1e01b31df43bbba4cd98ea4ac&vid=05a6dd2d7ca5ffe1e01b31df43bbba4cd98ea4ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR