Pyridine-2,3-dicarboxamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

P1215-25G P1215-5G
TCP1215-25GEA 799.95 CAD
TCP1215-25G TCP1215-005G
Pyridine-2,3-dicarboxamide ≥98.0% (by HPLC, total nitrogen)
Pyridine-2,3-dicarboxamide

CAS Number: 4663-94-9
MDL Number: MFCD00191488
Molecular Formula: C7H7N3O2
Molecular Weight: 165.15
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White

Formula: C₇H₇N₃O₂
MDL Number: MFCD00191488
CAS Number: 4663-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08bfed629056cc02311bce904cb20de27505dc08&vid=08bfed629056cc02311bce904cb20de27505dc08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR