3-Pentadecylphenol ≥90.0%

Supplier: TCI America

P1202-25G P1202-100G
TCP1202-025GEA 52.14 CAD
TCP1202-025G TCP1202-100G
3-Pentadecylphenol ≥90.0%
3-Pentadecylphenol

CAS Number: 501-24-6
MDL Number: MFCD00002310
Molecular Formula: C21H36O
Molecular Weight: 304.52
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 232
Melting point (°C): 50
Flash Point (°C): 113

Formula: C₂₁H₃₆O
Boiling Pt: 232 °C
Melting Pt: 50 °C
Flash Pt: 113 °C
MDL Number: MFCD00002310
CAS Number: 501-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f0b15167c12cdbe0360d60302399b0bdc198e2&vid=e9f0b15167c12cdbe0360d60302399b0bdc198e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR