Propyl-3-propoxypyridine-2-carboxylate ≥98.0%
Supplier: TCI America
P1196-5G
P1196-1G
TCP1196-5GEA
730.59
CAD
TCP1196-5G
TCP1196-001G
Propyl-3-propoxypyridine-2-carboxylate ≥98.0%
Propyl 3-propoxypyridine-2-carboxylate
CAS Number: 134319-22-5
MDL Number: MFCD00191575
Molecular Formula: C12H17NO3
Molecular Weight: 223.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 3-Propoxypicolinic Acid Propyl Ester, Propyl 3-Propoxypicolinate, 3-Propoxypyridine-2-carboxylic Acid Propyl Ester
Formula:
C₁₂H₁₇NO₃ Density: 1.07 (20 °C) |
MDL Number:
MFCD00191575 CAS Number: 134319-22-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...