N-(2-Pyridylmethyl)glycine ethyl ester ≥95.0%

Supplier: TCI America

Synonyms: N-(2-Picolyl)glycine Ethyl Ester, Ethyl N-(2-Pyridylmethyl)glycinate, Ethyl 2-((pyridin-2-ylmethyl)amino)acetate

P1194-5G P1194-1G
TCP1194-005GEA 395 CAD
TCP1194-005G TCP1194-001G
N-(2-Pyridylmethyl)glycine ethyl ester ≥95.0%
N-(2-Pyridylmethyl)glycine ethyl ester

CAS Number: 62402-24-8
MDL Number: MFCD00191573
Molecular Formula: C10H14N2O2
Molecular Weight: 194.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.10


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl N-(2-Pyridylmethyl)glycinate, N-(2-Picolyl)glycine Ethyl Ester

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Density: 1.10 (20 °C)
MDL Number: MFCD00191573
CAS Number: 62402-24-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95aeba8d27e5023cf521e5148427676e038c6641&vid=95aeba8d27e5023cf521e5148427676e038c6641&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR