O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime ≥98.0%

Supplier: TCI America

P1190-1G P1190-100MG P1190-5G
TCP1190-001GEA 532.46 CAD
TCP1190-001G TCP1190-100MG TCP1190-5G
O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime ≥98.0%
O-(2,3,4,5,6-Pentafluorobenzyl)formaldoxime

CAS Number: 86356-73-2
MDL Number: MFCD00191477
Molecular Formula: C8H4F5NO
Molecular Weight: 225.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Freezing point (°C): 24

Formula: C₈H₄F₅NO
MW: 225.12 g/mol
MDL Number: MFCD00191477
CAS Number: 86356-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c02f674228e30b33cb1e5fe9443ee832a2657c46&vid=c02f674228e30b33cb1e5fe9443ee832a2657c46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR