5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine ≥98.0%

Supplier: TCI America

Synonyms: 5-Acetyl-3-chloro-10,11-dihydro-5H-dibenz[b,f]azepine, 5-Acetyl-3-chloro-2,2'-iminodibenzyl

A2093-25G A2093-5G
TCA2093-25GEA 135.88 CAD
TCA2093-25G TCA2093-5G
5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine ≥98.0%
5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine

CAS Number: 25961-11-9
MDL Number: MFCD01632170
Molecular Formula: C16H14ClNO
Molecular Weight: 271.74
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 124


Other Product Names / Synonyms: 5-Acetyl-3-chloro-2,2'-iminodibenzyl

Formula: C₁₆H₁₄ClNO
MW: 271.74 g/mol
Melting Pt: 124 °C
MDL Number: MFCD01632170
CAS Number: 25961-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=555e0af86985885e023bb2eca4ab469b9d88b56d&vid=555e0af86985885e023bb2eca4ab469b9d88b56d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR