4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione

Supplier: TCI America

P1184-5G P1184-1G
TCP1184-5GEA 606.56 CAD
TCP1184-5G TCP1184-001G
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione
4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione

CAS Number: 4233-33-4
MDL Number: MFCD00003148
Molecular Formula: C8H5N3O2
Molecular Weight: 175.15
Form: Crystal
Color: Red
Storage Temperature: <0°C


Other Product Names / Synonyms: PTAD

Formula: C₈H₅N₃O₂
MDL Number: MFCD00003148
CAS Number: 4233-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b49364b3b5a5ea966ed66f14899dcfca85aad81&vid=6b49364b3b5a5ea966ed66f14899dcfca85aad81&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR